Learn More
United Scientific™ Beakers, New Style, PP
SDPSCI_ED
Catalog No. S00615 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
5000 mL
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S00615 25 mL
S00616 50 mL
S00617 100 mL
S00618 250 mL
S00619 500 mL
S00620 1000 mL
S00621 2000 mL
S00622 5000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. S00615 Supplier United Scientific™ Supplier No. 11110
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Large pouring spouts enable users to safely and accurately pour from these New Style PP Beakers.
  • They are autoclavable and feature raised graduations on the outer surface.
  • New Style beakers feature oversized, tapered spouts for easier pouring of solutions.
  • Beakers, molded in polypropylene, have excellent clarity and very good chemical resistance.
Specifications
Color Clear
Capacity (Metric) 25 mL
Material Polypropylene
Quantity 12 Pk.
Type Wide Spout Beaker
Autoclavable Autoclavable
Disposable Reusable
Graduated Yes
Scale Raised Outer Graduated Scale
Style Tapered