Learn More
Fluotics Beckman iSeries/FX/NX Compatible P80, non-sterile
Supplier:  Fluotics BFXI80.NS
Encompass
Beckman iSeries/FX/NX Compatible P80 non-sterile Working Volume 80uL Tip Color Clear Tip Box 96 Multi Channel Head Compare To 717254 B85764 50 racks/case with 96 tips per rack
Catalog No. 50-004-35254