Learn More
MilliporeSigma™ Beclin-1 Activator I, TAT-Beclin-1, Calbiochem™,
Catalog No. 5060480001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
Catalog No. Quantity
50-604-80001 10 mg
1 options
Catalog No. 50-604-80001 Supplier MilliporeSigma™ Supplier No. 5060480001
Add to Cart
Add to Cart
A cell-permeable Atg6/Beclin1-derived autophagy-inducing peptide, Peptide Sequence: H-Tyr-Gly-Arg-Lys-Lys-Arg-Arg-Gln-Arg-Arg-Arg-Gly-Gly-Thr-Asn-Val-Phe-Asn-Ala-Thr-Phe-Glu-Ile-Trp-His-Asp-Gly-Glu-Phe-Gly-Thr-OH; Packaged under inert gas
TRUSTED_SUSTAINABILITY
Specifications
Quantity 10 mg
Synonym H2N-YGRKKRRQRRR-GG-TNVFNATFEIWHDGEFGT-CO2H; Atg6 Activator I; Beclin1-GAPR-1 Interaction Blocker I; Vps30 Activator I; Autophagy Inducer IV