Learn More
Apexbio Technology LLC Bedaquiline 843663-66-1 10mg
SDP
Supplier:  Apexbio Technology LLC B349210
Encompass_Preferred
Bedaquiline a diarylquinoline antibiotic targets Mycobacterium tuberculosis via dual inhibition of subunits c and within the bacterial F1FO-ATP synthase complex disrupting ATP production It is applied primarily in research addressing multi-drug resistant (MDR) tuberculosis Recent studies have expanded its use into oncology where bedaquiline inhibits mitochondrial respiratory function and reduces glycolytic activity in cancer stem-cell-like populations In vitro experiments employing breast cancer MCF7 cells revealed bedaquiline increased ROS levels and reduced mitochondrial membrane potential Reported IC50 values for growth inhibition range approximately 1-10 M depending on cell lines and experimental conditions
Catalog No. 50-000-62699
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.