Learn More
Apexbio Technology LLC Bedaquiline fumarate 845533-86-0 5mg
SDP
Supplier:  Apexbio Technology LLC B34915
Encompass_Preferred
Bedaquiline fumarate (CAS 845533-86-0) is a small-molecule inhibitor targeting ATP synthase of Mycobacterium tuberculosis It is designed to inhibit mycobacterial ATP synthase thereby disrupting ATP biosynthesis and bacterial energy metabolism Bedaquiline fumarate exerts its biological activity primarily through selective inhibition of ATP synthase In biochemical assays bedaquiline fumarate demonstrates potent inhibitory activity with an IC50 value of approximately 30 nM against M tuberculosis ATP synthase Based on these pharmacological properties bedaquiline fumarate holds research potential in studying tuberculosis pathogenesis mechanisms of antibiotic resistance and evaluating therapeutic strategies for mycobacterial infections
Catalog No. 50-000-94501
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.