Learn More
MP Biomedicals™ Beef Extract, MP Biomedicals™
Catalog No. MP210088320 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
20 g
Catalog No. Quantity
MP210088320 20 g
1 options
Catalog No. MP210088320 Supplier MP Biomedicals™ Supplier No. 0210088320
Add to Cart
Add to Cart
Standardized for use in microbiological media. Beef Extract, MP Biomedicals is a water soluble powder, suitable for general purpose media. It is recommended for use in bacteriological examination of water, milk and other materials and used in biochemical
Beef extract is recommended for use in bacteriological examination of water, milk, and other materials, where uniform composition of media is important. It is also used for biochemical studies, particularly fermentation reactions, because of its independence from fermentable substances.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 20 g
WARNING: Reproductive Harm - www.P65Warnings.ca.gov