Learn More
Waters Corp ACQUITY UPLC BEH C18 Column, 130Å, 1.7 µm, 3 mm X 150 mm, 1/pk
Supplier:  Waters Corp 186004690
Encompass_Preferred
As the premier column for method development, the 1.7 um ACQUITY UPLC BEH C18 sorbent provides you with the most universal column choice for UPLC separations. The trifunctionally bonded BEH particle gives you the widest usable pH range (pH 1-12), superior low pH stability, and ultra-low column bleed for high sensitivity MS applications.
Catalog No. NC0754419
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.