Learn More
Description
- Covers to be used with Bel-Art™ 96 Deep-Well Plates (F378600001, F378600000)
- Specially designed polypropylene (PP), keyed fit cover separates each well with a sealing ring to reduce evaporation and permits safe and stable stacking of plates
- Covers are keyed to fit only one way
Specifications
Specifications
| Barcode | No Barcode |
| Array | 8 x 12 |
| Autoclavable | Autoclavable |
| Color | Natural |
| Product Type | Well Plate Lid |
| For Use With (Equipment) | SP Scienceware 96 Deep-Well Plates (F378600001, F378600000) |
| Material | Polypropylene |
| No. of Wells | 96 |
| Sterility | Non-sterile |
| Surface Treatment | Untreated |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.