Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Acid Pump/Reagent Dispenser
SCI_ED

Catalog No. S41365 Shop All SP Bel-Art & SP Wilmad-LabGlass Products
Change view
Click to view available options
Autoclavable:
Not Autoclavable
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Autoclavable
S41365 Not Autoclavable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S41365 Supplier Bel-Art™ Supplier No. F116560000/EMD
This item is not returnable. View return policy

Flow control for dangerous liquids.

Bottle provides pressure for delivery of steady liquid

Fits on standard 2.4L (5 pt) acid bottles or 3.8L (1 gal) chemical bottles. Relief valve button on top stops liquid flow from spout. Siphon tube long enough to reach bottom of 2.4L acid bottle.

Specifications

Angle 45 degrees
Product Type Reagent Dispenser
Autoclavable Not Autoclavable
Color White
Channels 1
Material Polyethylene
For Use With (Application) Dispense dangerous liquids without pouring
For Use With (Equipment) 2.5, 4L, 5 pt., 0.5 gal., 1 gal. jugs and bottles
Includes Two adapters for short neck bottles
Warranty No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.