Learn More
Bel-Art™ SP Scienceware™ Barbed Fittings with Reducing Connector
Shop All SP Bel-Art & SP Wilmad-LabGlass ProductsDescription
- Chemical-resistant polypropylene plastic for use with wide range of liquids or gases
- FDA-grade for direct contact with food, drugs, and cosmetics
- Autoclavable
Specifications
Specifications
| Autoclavable | Autoclavable |
| Material | Polypropylene |
| For Use With | 4.7 to 9.5mm |
| Tubing Size | For 3/16 in. to 1/2 in. tubing (4.7 to 9.5mm) |
| Includes | Reducing connector |
| Color | Clear |
| Product Type | Barbed Fitting |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.