Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Bel-Blotter™ 96-Well Replicating Tool

Catalog No. 1371213 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Volume (Metric):
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
13-712-13 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-712-13 Supplier Bel-Art™ Supplier No. F378760002
Get Up to $500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed to do tedious tasks with a minimum of effort

  • The 96 open-ended pipette tips of this tool fit all types of 96 well plates from flat, vee or round bottom plates to 0.2mL thin walled PCR plates and tubes
  • Can be used for replication of recombinant DNA libraries, inoculation of filters for colony hybridization, PCR, phage typing and other applications
  • Made of polycarbonate plastic, it is easy-to-use, reusable and autoclavable

Specifications

Autoclavable Autoclavable
Material Polycarbonate
Sterility Non-sterile
For Use With (Application) Increase Productivity, Minimize Effort
Quantity 1 Pack
Color Transparent
Volume (Metric) 0.2 mL
Length (Metric) 12.8 cm
Type 96-Well Replicating Tool
Disposable Reusable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.