Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Sterileware™ Bent Handle Scoop

Catalog No. 14241203 Shop All SP Bel-Art & SP Wilmad-LabGlass Products
Change view
Click to view available options
Capacity (Metric):
60 mL
125 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
14-241-203 125 mL
14-241-202 60 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 14-241-203 Supplier Bel-Art™ Supplier No. H369042006
This item is not returnable. View return policy

Features an ergonomic, curved handle design to elevate the hand from the sample, reducing risk of contamination

  • Curved handle helps prevent incidental contact with sample source material and exposure to hazardous substances
  • Flat bottom keeps scoop upright when placed on a flat surface; can be used as a weigh boat
  • Made from high-impact, lightweight, FDA grade polystyrene
  • Sterilized by gamma irradiation, lot stamped for accurate lot tracing, and shipped with a Certificate of Sterilization
  • Individually packaged and ready for one time use and disposal

Specifications

Sterility Sterile
Autoclavable Not Autoclavable
Capacity (English) 4 oz.
Capacity (Metric) 125 mL
Certifications/Compliance FDA
Disposable Single-use
Product Type Scoop
For Use With (Application) Design to elevate the hand from the sample, reducing risk of contamination, prevent incidental contact with sample source material and exposure to hazardous substances
Length (English) 7.62 in.
Width (English) 2 in.
Width (Metric) 5.1 cm
Dimensions (L x W) 7.68 x 2 x 1.31 in. (19.4 x 5.1 x 3.3 cm)
Includes Certificate of processing for sterilization
Length (English) Handle 3.87 in.
Length (Metric) Handle 9.8 cm
Material Polystyrene
Handle Type Bent, Curved
Length (Metric) 19.4 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.