Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Cleanware™ Bottle Brush
GSA_VASCI_ED

Catalog No. S90135 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Color:
Natural
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
S90135 Natural
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S90135 Supplier Bel-Art™ Supplier No. F170680000/EMD

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Uniquely designed, swiveling pivot head enables thorough cleaning of bottles and flasks including difficult curves and corners

  • Thoroughly cleans glass and plastic labware without scratching
  • Swivel head fits through a 29/42 standard taper neck, or any non-tapered neck with a diameter of 24mm (1 in.) or larger
  • Brush head is slightly convex, 45 x 21mm (L x W) with bristles 8mm long
  • Brush head is attached to hardwood handle 37cm long x 7mm diameter
  • Features natural bristles, swivel head and small brush head

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Product Type Bottle Brush
Bristle Material Natural
Bristle Type Stiff
Color-coded No
Handle Material Wood
Handle Type Round
Tip Type Bent
Scent Unscented
Sterility Non-sterile
Color Natural
For Use With (Application) Use this brush to clean any type of deep vessel, good for restaurant kitchens, laboratories with round-bottom flasks, repair shops, any facility needing dexterity and small size in the cleaning head
Length (English) Handle 13 in.
Length (Metric) Handle 33 cm
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.