Description
- One piece
- Made from bonded neoprene
- For glove boxes with 6 in. glove ports
Specifications
Specifications
| Color | Black |
| Hand | Specific |
| Latex Free | No |
| Material | Bonded Neoprene |
| Product Type | Glove Box Gloves |
| Size | 11 |