Learn More
Bel-Art™ SP Scienceware™ Glove Box Clamping Rings
Adjustable rings hold gloves in place
$57.10
Specifications
Product Type Clamping Ring
For Use With (Application) Fasten Glove Firmly to Glove Box
View More Specs
Products 2
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
19-821-983
View Documents
Bel-Art™
H500250320
Fits 8 in. Circular Port Holes
Each for $57.10
Only null left
Add to Cart
03-421-102
View Documents Promotion Details
Bel-Art™
H500290200
Fits 6 in. Circular Port Holes N/A N/A N/A
Description
Description
  • Fit Techni-Dome™ glove boxes(Mfr. No. 500300000) or other glove boxes with circular port holes of the correct diameter
Specifications
Specifications
Clamping Ring
Fasten Glove Firmly to Glove Box
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.