Learn More
Bel-Art™ Heavy Duty Pail
Catalog No. 22163659 Shop All SP Bel-Art & SP Wilmad-LabGlass Products
Catalog No. 22-163659
Supplier: Bel-Art™ H168060000
Only null left
Add to Cart
Add to Cart
Polyethylene pails have wide rims that hold their shape under heavy loads
  • Features include a heavy-gauge, plastic-coated metal bail and a pronounced spout
  • Recessed hand grip on the bottom aids in safe handling and pouring.
  • Graduations are in liters and molded on the interior
Specifications
Product Type Heavy Duty Pail
Graduations Liters
Material Polyethylene
Color White
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.