Learn More
Bel-Art™ Octagon Magnetic Stirring Bar Assortments
Catalog No. 22331470 Shop All SP Bel-Art & SP Wilmad-LabGlass Products
Change view
Click to view available options
Quantity:
12 std bars (2 ea. size)
12 std bars (2 ea. size) + 2 micro
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-331470 12 std bars (2 ea. size) + 2 micro
22-331462 12 std bars (2 ea. size)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-331470 Supplier Bel-Art™ Supplier No. F371800000
Add to Cart
Add to Cart
Popular sizes and shapes
  • Convenient storage box segregates sizes and styles for easy, no-rummage organization
  • Octagonal spin bars, each with pivot ring
  • Made from Teflon™ PTFE
  • Autoclavable
Specifications
Length (Metric) 12.7 mm, 15.7 mm, 25.4 mm, 38.1 mm, 50.8 mm, 63.5 mm
Diameter (Metric) 7.9 mm, 9.4 mm
Quantity 12 std bars (2 ea. size) + 2 micro
Coating PTFE, FDA-grade, USP Class VI
Material PTFE, Alnico Magnet
Type Magnetic Stir Bar Set
Shape Octagon
Color White
Certifications/Compliance FDA
Includes 2 micro and 12 standard bars two each of 0.5 x 0.31 in., 0.62 x 0.31 in., 1 x 0.37 in., 1.5 x 0.37 in., 2 x 0.37 in., 2.5 x 0.37 in.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov