Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel Art™ Poxygrid™ Wire Test Tube Racks, Holds Tubes 10 to 13 mm

Catalog No. 22260540 Shop All SP Bel-Art & SP Wilmad-LabGlass Products
Change view
Click to view available options
Color:
Blue
Green
Orange
Positions:
60
72
108
150
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Positions
22-260540 Green 72
22-260541 Blue 72
22-260545 Blue 108
14-793-231 Blue 60
14-793-235 Blue 150
14-512-03 Orange 72
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 22-260540 Supplier Bel Art™ Supplier No. F187500000
This item is not returnable. View return policy

Chemical (organic solvents, salts, weak acids and weak alkalis) and corrosion-resistant steel wire racks are coated with heat-cured epoxy

  • Withstands temperature range of -223°C (-369°F) to 121°C (250°F)
  • Consists of square holes, three decks, a slotted grid bottom and extra strength reinforced edges
  • Orange racks can be used for biohazard warning while yellow for radioactive warning
  • Epoxy coatings are unaffected by organic solvents, salts, weak acids and weak alkalis
  • Stores from 40 to 108 tubes (five different diameters of testtubes)
  • Autoclavable

Specifications

Autoclavable Autoclavable
Color Green
Holds Tubes (10 to 13 mm)
Material Epoxy-coated Steel
Positions 72
Shape Rectangular
Stackable Yes, When Empty
Sterility Non-sterile
Dimensions (L x W x H) 8 x 4.4 x 2.5 in. (20.5 x 11.3 x 6.4cm)
Length (Metric) 20.5 cm
Length (English) 8.07 in.
Width (English) 4.45 in.
Height (English) 2.51 in.
Diameter (Metric) Opening 13 mm
Product Type Test Tube Rack
Shelves None
Height (Metric) 6.4 cm
Array 6 x 15
Width (Metric) 11.3 cm
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.