Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Reagent/Acid Pump Dispenser
GREENER_CHOICE

Catalog No. 01002 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Autoclavable:
Not Autoclavable
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Autoclavable
01-002 Not Autoclavable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 01-002 Supplier Bel-Art™ Supplier No. F116560000
Get Up to $500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Aids in the transfer of acids and reagents and virtually eliminates accidental bottle breakage and spills

  • Squeeze the small plastic bottle to begin flow; press the pressure release button to immediately stop dispensing
  • Fits many 5 pint (80 fl. oz.) acid bottles, 1/2 and 1 gallon jugs, 2.5 and 4 liter bottles, and similar bottles with GPS No. 38-430 or 38-400 threaded necks (not included)
  • Two adapters provided for short necks

Specifications

Angle 45 degrees
Product Type Reagent Dispenser
Autoclavable Not Autoclavable
Color White
Channels 1
Material Polyethylene
Volume (Metric) 4 L
For Use With (Application) Dispense dangerous liquids without pouring
For Use With (Equipment) 2.5, 4L, 5 pt., 0.5 gal., 1 gal. jugs and bottles
Includes Siphon tube long enough to reach bottom of 2.4L Acid bottles
Warranty No
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.