Learn More
Bel Art™ SP Scienceware™ Reusable Desiccant Cartridges
Convenient, easy-to-handle cartridges with Natrasorb™ TI indicating silica gel beads. Bel Art SP Scienceware™ Reusable Desiccant Cartridges can also be used with other plastic desiccators of similar size.
Specifications
Product Type Scienceware™ Reusable Desiccant Cartridge
Material Polyester
View More Specs
Description
Description
  • Desiccant changes color from blue when dry to pink when moist Saturated gel can be regenerated in a 150°C (300°F) vented oven until blue color returns
  • Cartridge can be regenerated repeatedly without degradation
  • Desiccant beads contained in a durable, polyester felt bag with a perforated Mylar°Cover Cartridges are individually sealed in moisture proof pouches ready for immediate use
Order Info
Fits Bel-Art Space Saver™ desiccators in addition to other desiccators of similar size.
Specifications
Specifications
Scienceware™ Reusable Desiccant Cartridge
Polyester
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.