Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Sterileware™ Bent Handle Scoops
SCI_ED

Catalog No. s04787 Shop All SP Bel-Art & SP Wilmad-LabGlass Products
Change view
Click to view available options
Capacity (Metric):
60 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S04787 60 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S04787 Supplier Bel-Art™ Supplier No. H369022006/EMD
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Features an ergonomic, curved handle design to elevate hand from the sample, reducing risk of contamination and minimizing wrist strain.

  • Made from high-impact, lightweight, FDA grade polystyrene
  • Curved handle helps prevent incidental contact with sample source material and exposure to hazardous substances
  • Flat bottom keeps scoop upright when placed on flat surface, can be used as weigh boat
  • Sterilized by gamma irradiation, lot stamped for accurate lot tracing, and shipped with Certificate of Sterilization
  • Individually packaged and ready for one time use and disposal
  • Pack of 10

Specifications

Sterility Sterile
Autoclavable Not Autoclavable
Capacity (English) 2 oz.
Capacity (Metric) 60 mL
Certifications/Compliance FDA
Disposable Single-use
Product Type Scoop
For Use With (Application) Design to elevate the hand from the sample, reducing risk of contamination, prevent incidental contact with sample source material and exposure to hazardous substances
Length (English) 7.5 in.
Width (English) 1.62 in.
Width (Metric) 4.1 cm
Dimensions (L x W) 7.5 x 1.62 x 1.5 in. (19.1 x 4.1 x 3.7 cm)
Includes Certificate of processing for sterilization
Length (English) Handle 4.56 in.
Length (Metric) Handle 11.6 cm
Material Polystyrene
Handle Type Bent, Curved
Length (Metric) 19.1 cm
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.