Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Super Biohazard Disposal Bags
SCI_ED

Catalog No. S90149A Shop All SP Bel-Art & SP Wilmad-LabGlass Products
Change view
Click to view available options
Dimensions (L x W):
19 x 14 in. (48 x 36 cm)
23 x 19 in. (58 x 48 cm)
35 x 25 in. (35 x 64 cm)
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
S90149A 19 x 14 in. (48 x 36 cm)
S90149C 35 x 25 in. (35 x 64 cm)
S90149B 23 x 19 in. (58 x 48 cm)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S90149A missing translation for 'mfr' Bel-Art™ missing translation for 'supplierNo' F131651419

Protects against dangerous tears and leaks. Super Biohazard Disposal Bags feature a sterilization indicator patch that darkens when exposed to steam sterilization for 20 minutes.

  • Protect against dangerous tears and leaks
  • Orange-red bags have standard biohazard warning symbol and precautionary procedures in four languages: English, French, German, and Spanish
  • Extra-heavy-duty bags are 2 mil-thick blended polymer
  • Autoclavable to 121°C (250°F)
  • Sterilization indicator patch darkens when exposed to steam sterilization

Specifications

Capacity (Metric) 165 L
Closure Type Zip Tie
Capacity (English) 43.6 gal.
Color Red
Dimensions (L x W) 19 x 14 in. (48 x 36 cm)
Disposable Single-use
Length (Metric) 48 cm
Sterility Non-sterile
Width (Metric) 36 cm
Autoclavable Autoclavable
Includes Standard biohazard warning symbol and precautionary procedures in four languages: English, French, German and Spanish
For Use With (Application) Extra-strength, high-impact, high-temperature bags reduce autoclave time
Width (English) 14 in.
Length (English) 19 in.
Thickness 2 mil
Material Polymer
Product Type Biohazard Disposal Bag
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.