Learn More
Selleck Chemical LLC Belinostat (PXD101) 200mg 414864-00-9 NSC726630, PX-105684
SDP
Supplier:  Selleck Chemical LLC S1085200MG
Encompass_Preferred
Belinostat (PXD101, NSC726630, PX-105684) is a novel HDAC inhibitor with IC50 of 27 nM in a cell-free assay, with activity demonstrated in cisplatin-resistant tumors. Belinostat (PXD101) induces autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-835-4
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.