Learn More
Cytiva Benchkote sheet for ÄKTA systems
Catalog No. 09920056 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
10/Pk.
25/Pk.
50/Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-920-056 25/Pk.
09-920-054 10/Pk.
09-920-055 50/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 09-920-056 Supplier Cytiva Supplier No. 230010094
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart
Benchkote sheets for ÄKTA systems will protect the top buffer trays from buffer spillages and salt deposits.
Why Benchkote sheet for lab ÄKTA systems?
  • Cover the top buffer tray with Benchkote sheet and protect from buffer spillages and salt deposits.
  • Simplify daily and weekly preventive maintenance of the top buffer tray of your ÄKTA systems
  • Smooth, absorbent Whatman™ paper quickly absorbs spills.
  • Laminated polyethylene layer prevents flow-through to the working surface.
  • Offered in three convenient pack sizes: 10/pk, 25/pk and 50/pk sheets.
  • Convenient and easy to use.
  • Benchkote sheets are manufactured of absorbent, impermeable material designed to protect lab surfaces against hazardous spills.
  • The high-quality, smooth, and absorbent Whatman™ paper, quickly absorbs liquid spills, and laminated polyethylene layer prevents flow-through to the working surface.
TRUSTED_SUSTAINABILITY
Specifications
Material Absorbent Paper, Laminated Polyethylene
Description For use with ÄKTA go
Quantity 25/Pk.