Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Benchtop Extraction Kit
GREENER_CHOICE

Catalog No. 15050088 Shop All Fisherbrand Products
Change view
Click to view available options
Product Type:
Carbon Extraction Kit
Carbon Filter
Filter Connector
Standard Extraction Kit
Standard Filter
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
15-050-088 Standard Filter
15-050-086 Standard Extraction Kit
15-050-087 Carbon Extraction Kit
15-050-089 Carbon Filter
15-050-090 Filter Connector
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 15-050-088
Add to Cart
Add to Cart
This item is not returnable. View return policy

Benchtop Extraction Kits used for the capture and containment of particulate generated on the bench space.

  • The Benchtop extraction kits include arms with hood or nozzle, fan with speed control and standard filter (particle, HEPA or gas), hose, connection details, and table bracket.
  • The speed of the fan is variably controlled by a speed controller.
  • Complete with hoses and extraction hoods.
  • The hood is made of clear plastic, which increases the visibility.

Specifications

Product Type Standard Filter
Color White
Material Aluminum
For Use With (Equipment) Benchtop Extraction Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.