Learn More
Description
- The Benchtop extraction kits include arms with hood or nozzle, fan with speed control and standard filter (particle, HEPA or gas), hose, connection details, and table bracket.
- The speed of the fan is variably controlled by a speed controller.
- Complete with hoses and extraction hoods.
- The hood is made of clear plastic, which increases the visibility.
Specifications
Specifications
| Product Type | Carbon Filter |
| Color | White |
| Material | Aluminum |
| For Use With (Equipment) | Benchtop Extraction Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.