Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sartorius Benchtop Meters: pHBasic+ Series

Catalog No. 14558068 Shop All Sartorius Products
Change view
Click to view available options
Type:
pH Meter with Kit
pH Meter with Kit and Electrode
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
14-558-068 pH Meter with Kit
14-558-069 pH Meter with Kit and Electrode
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-558-068 Supplier Sartorius Supplier No. PHBASIC(PLUS)X

For entire pH requirements

  • Fast and accurate results, easy operation, and capacity to have direct data recall and transfer
  • Full functionality with USB data transmission
  • Recall and output of 5,000 data set memory
  • Easy-to-use menus and intuitive operation sets
  • Sensor efficiency & calibration integrity
  • Clear status icons and indicators
  • Large LCD display
  • Automatic temperature compensation
  • Automatic buffer recognition
  • Programmable data collection
  • 5-point calibration gives a broader range in slope accuracy
  • Features Continuous Monitoring Control CMC, GLP TIME/DATE stamps
TRUSTED_SUSTAINABILITY

Specifications

Description Meter, Power Supply, Cover, Arm (5pt Cal, ElectCheck, USB,CMC)
Outputs USB
Includes Power Supply, Manual, 4 AA batteries, Electrode Arm and Base
For Use With (Application) Sample and Buffer Preparation - Regulated R&D and QC environments
Calibration Points 5
Display Type LCD
Memory Data Memory 500 and output
Warranty 2 Years
Model pHBasic+
Type pH Meter with Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.