Learn More
Sartorius Benchtop Meters: Pro 15 Series
Catalog No. 14558074 Shop All Sartorius Products
Change view
Click to view available options
Calibration Points:
5
Catalog No. Calibration Points
14-558-074 5
1 options
Catalog No. 14-558-074 Supplier Sartorius Supplier No. PR15BIOKIT
For advanced professional pH and ISE analyses
  • Exact measurements with wider measuring ranges and better accuracy specifications
  • Advance GLP Documentation with time and date stamps
  • Direct data transmission with RS232 and USB interfaces
  • Automated calibration with up to 5 points Automatic buffer recognition out of 22 buffer sets
  • Multi-functionality in One
  • Complete Documentation
  • Reliable results with automatic and wireless sensor and user ID recognition
  • User name can be assigned to each measured value
  • Complies with GLP requirements with administrator rights or user identification via electronic ID card
  • Large LCD display, Intuitive menu navigation, ElectCheck and Stability Control
  • Offers highest level of complete compliance in GLP documentation with direct data transmission 1500 data
TRUSTED_SUSTAINABILITY
Specifications
Outputs USB, RS-232
Calibration Points 5
Description Meter, B&W backlit display, Power supply, Cover, Arm, 3 Buffers, Manual, Electrode PY-P28-2S (5pt,GLP)
Display Type LCD
For Use With (Application) Sample and Buffer Preparation - FDA and UPS Regulated R&D and QC environments - Biological environments: Cell culture, blood separation, microbiology, proteomics genomics, and molecular biology
Includes Power Supply, Manual, 4 AA batteries, Electrode Arm and Base, Electrode PY-P28-2S
Memory Data Memory 1500 and output
Product Type PR-15 Bio Kit with PY-P28-2S electrode
Warranty 2 Years