Learn More
Sartorius Benchtop Meters: Pro 50 Series
Catalog No. 14558078 Shop All Sartorius Products
Change view
Click to view available options
Calibration Points (pH):
5
Catalog No. Calibration Points (pH)
14-558-078 5
1 options
Catalog No. 14-558-078 Supplier Sartorius Supplier No. PR50BIOKIT
For advanced professional pH, ISE, DO, Conductivity analyses
  • Exact measurements with wider measuring ranges and better accuracy specifications
  • Advance GLP Documentation with time and date stamps
  • Direct data transmission with RS232 and USB interfaces
  • Automated calibration with up to 5 points Automatic buffer recognition out of 22 buffer sets
  • Multi-functionality in One
  • Complete Documentation
  • Reliable results with automatic and wireless sensor and user ID recognition
  • User name can be assigned to each measured value
  • Complies with GLP requirements with administrator rights or user identification via electronic ID card
  • Large LCD display, Intuitive menu navigation, ElectCheck and Stability Control
  • Offers highest level of complete compliance in GLP documentation with direct data transmission 1500 data
TRUSTED_SUSTAINABILITY
Specifications
Outputs USB and RS232 Interface
Calibration Points (pH) 5
Description Meter with ID sensor Card. B&W Display. Menu Structured Handling. Recorder Function. pH, mV, Temp, ISE., Conductivity, DO RS232- USB slave 1BNC . Power Supply. Cover, Arm,3 Buffers,Electrode PY-P28-2S (5pt,GLP)
Display Type Large LCD
For Use With (Application) Sample and Buffer Preparation FDA and UPS Regulated R&D and QC environments - Biological environments: Cell culture, blood separation, microbiology, proteomics genomics, and molecular biology
Includes Power Supply, Manual, 4 AA batteries, Electrode Arm and Base, Electrode PY-P28-2S
Memory Data Memory 1500 and output
Product Type PR-50 Bio Kit with PY-P28-2S electrode
Warranty 2 Years