Learn More
Cole-Parmer Cole-Parmer Benchtop Splash-Guard, Arcylic, 24" W x 24" H x 1/4" Thick
Supplier: Cole-Parmer 0677110
Protect yourself from splashing water, flying debris and mishaps in the lab. This splash-guard is fabricated from 1/4" clear acrylic and is solvent bonded to a plastic base. Position several units along the benchtop to keep lab set-ups separated. Includes two finger holes for easy lifting and carrying.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.