Learn More
Poltex™ Benchtop Stack System Cubby
SDP
Catalog No. 22000797
Change view
Click to view available options
Height (Metric):
3 in.
4 in.
5 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (Metric)
22-000-797 4 in.
22-000-800 3 in.
22-000-811 5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-000-797 Supplier Poltex™ Supplier No. BTSYSCUBBY4
Add to Cart
Add to Cart
As part of the Benchtop Storage System, these products can stack on one another using our foot and pocket design to create ideal benchtop organization for your lab.
  • Super durable and cleanable PETG construction
  • Rubber bumpons on base for non-slip placement
Specifications
Product Type Lab Box Organizer
Color Clear
Height (Metric) 4 in.
Material PETG