Learn More
Description
Specifications
Specifications
| Gene ID (Entrez) | 139105 |
| Species | Human |
| Purity | >80% by SDS-PAGE and Coomassie blue staining |
| Common Name | BEN domain containing 2, BEN domain-containing protein 2, chromosome X open reading frame 20, CXorf20, MGC33653 |
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| Formulation | PBS and 1M Urea, pH 7.4. |
| For Use With (Application) | This Recombinant Antigen is Only Intended to be Used as a Blocking Agent to Confirm Antibody Specificity with The Corresponding Antibody, Catalog Number NBP2-53903. It is Purified by IMAC Chromatography, and The Expected Concentration is Greater Than 0.5 mg/ml |
| Gene Symbol | BEND2 |
| Molecular Weight (g/mol) | 33 kDa |
| Preservative | No Preservative |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
