Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BEND4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501734501 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-4501 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-4501 Supplier Proteintech Group Inc Supplier No. 247111AP150UL

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

BEND4, also named as CCDC4, has 5 isoforms with MW 55-58 kDa, 45-48 kDa and 36 kDa.

Specifications

Antigen BEND4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene BEND4
Gene Accession No. P86174, Q6ZU67
Gene Alias BEN domain containing 4, BEND4, CCDC4
Gene Symbols BEND4
Host Species Rabbit
Immunogen BEND4 Fusion Protein Ag20169
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 389206, 666938
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.