Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benedict's Solution, Quantitative, Ricca Chemical
SDP

Catalog No. 93516 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
93516 500 mL
R0935000100 100 mL
93532 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 93516 Supplier Ricca Chemical Company Supplier No. 93516
Add to Cart
Add to Cart

Specifications

Chemical Name or Material Benedict's Solution
Color Blue
Name Note Quantitative
CAS 7732-18-5, 63126-89-6, 333-20-0, 497-19-8, 7758-99-8, 14459-95-1
CAS Min % 68.0, 13.56, 8.48, 6.78, 1.22, 0.02
CAS Max % 71.0, 14.12, 8.82, 7.06, 1.27, 0.02
Grade Laboratory
Packaging Natural Poly Bottle
pH Alkaline
Quantity 500 mL
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.