Learn More
Apexbio Technology LLC Divalproex Sodium 76584-70-8 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B13915.1
Encompass_Preferred
Divalproex Sodium (CAS 76584-70-8) is a small-molecule inhibitor targeting histone deacetylases (HDACs) and modulating GABAergic neurotransmission It is designed to inhibit HDACs and modulate associated signaling pathways thereby regulating neuronal excitability and gene expression Divalproex Sodium exerts its biological activity primarily through HDAC inhibition enhancement of GABA levels inhibition of voltage-gated sodium channels and modulation of NMDA receptor-mediated glutamatergic signaling In experimental studies Divalproex Sodium demonstrates HDAC inhibitory activity with reported IC50 values typically ranging between 400 600 M varying by assay Based on these pharmacological properties Divalproex Sodium holds research potential in epilepsy bipolar disorder and migraine prophylaxis
Catalog No. 50-000-91198
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.