Learn More
Selleck Chemical LLC Benidipine HCl S2017-1g
SDP
Supplier:  Selleck Chemical LLC S20171G
Encompass_Preferred
Benidipine HCl (KW-3049 ( )- Benidipine) is a hydrochloride salt form of benidipine which is a dihydropyridine calcium channel blocker
Catalog No. 50-310-9655
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.