Learn More
Selleck Chemical LLC Chlormezanone S2021-50mg
SDP
Supplier:  Selleck Chemical LLC S202150MG
Encompass_Preferred
Chlormezanone a non-benzodiazepine that is used in the management of anxiety It has been suggested for use in the treatment of muscle spasm
Catalog No. 50-310-9656
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.