Learn More
TARGETMOL CHEMICALS INC PEMETREXED DISODIUM HEMI 500MG
Supplier: TARGETMOL CHEMICALS INC T6226500MG
Also available in 50 mg 100 mg 200 mg and bulk. Please contact Fisher for quotes. Pemetrexed disodium hemipenta hydrate (LY-231514 Disodium Hydrate) is a new-type antifolate and antimetabolite for TS DHFR and GARFT. The Ki of Pemetrexed Disodium Hydrate for TS DHFR and GARFT is 1.3 nM 7.2 nM and 65 nM respectively. purity: 99%
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.