Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bentazon, Restek
SDP

Catalog No. 06719882 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-719-882 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-719-882 Supplier Restek Corporation Supplier No. 32257

Chemical Identifiers

CAS 25057-89-0
Molecular Formula C10H12N2O3S
Molecular Weight (g/mol) 240.277
InChI Key ZOMSMJKLGFBRBS-UHFFFAOYSA-N
PubChem CID 2328
ChEBI CHEBI:3018
IUPAC Name 2,2-dioxo-3-propan-2-yl-1H-2$l^{6},1,3-benzothiadiazin-4-one
SMILES CC(C)N1C(=O)C2=CC=CC=C2NS1(=O)=O

Specifications

Quantity 1 mL
Concentration or Composition (by Analyte or Components) 1000ug/mL, Methanol
Chemical Name or Material Bentazon, Herbicide Mix
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.