Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benz[a]anthracene 98.0+%, TCI America™
SDP

Catalog No. B001725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B001725G 25 g
B00171G 1 g
B00175G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B001725G Supplier TCI America Supplier No. B001725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 56-55-3
Molecular Formula C18H12
Molecular Weight (g/mol) 228.29
MDL Number MFCD00003599
InChI Key DXBHBZVCASKNBY-UHFFFAOYSA-N
Synonym benz a anthracene, tetraphene, 1,2-benzanthracene, benzanthrene, benzo a anthracene, benzanthracene, naphthanthracene, 2,3-benzphenanthrene, benzoanthracene
PubChem CID 5954
ChEBI CHEBI:51348
IUPAC Name tetraphene
SMILES C1=CC=C2C=C3C(C=CC4=CC=CC=C34)=CC2=C1

Specifications

Melting Point 161°C
Boiling Point 438°C
Quantity 25 g
UN Number 3077
Formula Weight 228.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Benz[a]anthracene
RTECSNumber CV9275000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.