Learn More
Lkt Laboratories CAS 91080-29-4, ≥95%, Cationic surfactant used as a disinfectant. It dissociates bacterial lipid membranes, allowing cellular leakage. It is most effective against gram positive bacteria.
SDP
Supplier:  Lkt Laboratories B1545
Encompass_Preferred
Benzalkonium is a quaternary ammonium cationic surfactant that exhibits antimicrobial activity. Benzalkonium is clinically used as a disinfectant; it displays antibacterial activity by inducing dissociation of lipid membrane bilayer components and allowing cellular leakage. Benzalkonium is more active against gram positive bacteria but displays a wide spectrum of activity. This compound may also induce bronchoconstriction.
Catalog No. NC2539790
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.