Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Benzamidine Sepharose™ 4 Fast Flow, High Sub Media

Catalog No. 45000280 Shop All Cytiva Products
Change view
Click to view available options
Particle Size:
90 μm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Particle Size
45-000-280 90 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-000-280 Supplier Cytiva Supplier No. 17512310
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For convenient, one-step removal and/or purification of trypsin, trypsin-like serine proteases and thrombin

  • p-aminobenzamide (pABA) immobilized on Sepharose 4 Fast Flow matrix gives high productivity and allows easy scale-up
  • Binding capacity ≥35mg trypsin/mL media
  • Effective removal of thrombin and factor Xa after tag cleavage of recombinant proteins

TRUSTED_SUSTAINABILITY

Specifications

Particle Size 90 μm
pH 2 to 8 (long term); 1 to 9 (short term)
Description 25 mL
Material Benzamine Sepharose 4 Fast Flow
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.