Learn More
Sigma Organic Chemistry Benzene-1 3-dicarbonylo | 1G | MFCD29041544
Supplier: Sigma Organic Chemistry BOG002131G

Benzene-1 3-dicarbonylo , 1G
About this Item:
MDL #: MFCD29041544
Molecular Weight: 360.28
UNSPSC Code: 12352200
Chemical Formula: C16H12N2O8
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.