Learn More
Sigma Organic Chemistry Benzene-1 4-dithiocarbo | 1G | 13363-51-4 | MFCD00046866
Supplier: Sigma Organic Chemistry BOG001271G

Benzene-1 4-dithiocarbo , 1G
About this Item:
CAS #: 13363-51-4
MDL #: MFCD00046866
Molecular Weight: 196.29
UNSPSC Code: 12352200
Chemical Formula: C8H8N2S2
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.