Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzenepentacarboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. B095225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B095225G 25 g
B09525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B095225G Supplier TCI America Supplier No. B095225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1585-40-6
Molecular Formula C11H6O10
Molecular Weight (g/mol) 298.159
MDL Number MFCD00020281
InChI Key QNSOHXTZPUMONC-UHFFFAOYSA-N
PubChem CID 15314
IUPAC Name benzene-1,2,3,4,5-pentacarboxylic acid
SMILES C1=C(C(=C(C(=C1C(=O)O)C(=O)O)C(=O)O)C(=O)O)C(=O)O

Specifications

Color White
Quantity 25 g
Formula Weight 298.16
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Benzenepentacarboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.