Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzhydrylamine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. A0839250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0839250G 250 g
A083925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0839250G Supplier TCI America Supplier No. A0839250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5267-34-5
Molecular Formula C13H14ClN
Molecular Weight (g/mol) 219.712
MDL Number MFCD00013023
InChI Key CIHWJRSPVJBHGT-UHFFFAOYSA-N
Synonym benzhydrylamine hydrochloride, aminodiphenylmethane hydrochloride, benzhydrylamine, hydrochloride, diphenylmethanamine hydrochloride, benzhydrylammonium chloride, aminodiphenylmethane hcl, 1,1-diphenylmethylamine hydrochloride, diphenylmethylamine, chloride, benzhydrylamin hydrochloride, bha-polystyrene
PubChem CID 78921
IUPAC Name diphenylmethanamine;hydrochloride
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)N.Cl

Specifications

Melting Point 293°C
Color White
Quantity 250 g
Formula Weight 219.71
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Benzhydrylamine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.