Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzilic acid, 99%, Thermo Scientific™

Catalog No. p-7020878
Change view
Click to view available options
Quantity:
2500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA129830E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA129830E Supplier Thermo Scientific Chemicals Supplier No. A129830E

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Benzilic acid is used in organic synthesis, as a base point for preparation of glycollate pharmaceuticals and some hallucinogenic (deliriant) drugs - BZ, and Benactyzine being two examples. It is also used to determine zirconium and to get benzophenone.

Solubility
Sparingly soluble in water (1.4g/L at 25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-93-7
Molecular Formula C14H12O3
Molecular Weight (g/mol) 228.247
MDL Number MFCD00004447
InChI Key UKXSKSHDVLQNKG-UHFFFAOYSA-N
Synonym benzilic acid, diphenylglycolic acid, benzylic acid, hydroxy diphenyl acetic acid, diphenylhydroxyacetic acid, glycolic acid, diphenyl, hydroxydiphenylacetic acid, acide diphenylhydroxyacetique, benzilsaeure, alpha-hydroxy-2,2-diphenylacetic acid
PubChem CID 6463
ChEBI CHEBI:39414
IUPAC Name 2-hydroxy-2,2-diphenylacetic acid
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C(=O)O)O

Specifications

Melting Point 148°C to 152°C
Density 1.28
Boiling Point 180°C (13 mmHg)
Flash Point 215°C (419°F)
Odor Odorless
Quantity 2500 g
Beilstein 521402
Merck Index 14,1080
Solubility Information Sparingly soluble in water (1.4g/L at 25°C).
Formula Weight 228.25
Percent Purity 99%
Chemical Name or Material Benzilic acid
Show More Show Less
Hazard Category H302
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 200-993-2
RTECSNumber DD2064000
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.