Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzimidazole-6-carbonitrile, 97%, Thermo Scientific Chemicals

Catalog No. AAH6662303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6662303 1 g
AAH6662306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6662303 Supplier Thermo Scientific Chemicals Supplier No. H6662303

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6287-83-8
Molecular Formula C8H5N3
Molecular Weight (g/mol) 143.15
MDL Number MFCD06809988
InChI Key NICYTXJGZRYCEQ-UHFFFAOYSA-N
Synonym 1h-benzimidazole-5-carbonitrile, 1h-benzo d imidazole-6-carbonitrile, 1h-1,3-benzodiazole-5-carbonitrile, 1h-benzo d imidazole-5-carbonitrile, 5-cyanobenzimidazole, 1h-benzimidazole-6-carbonitrile, 5-benzimidazolecarbonitrile, benzimidazole-5-carbonitrile, 1h-benzoimidazole-5-carbonitrile, 3h-1,3-benzodiazole-5-carbonitrile
PubChem CID 223906
IUPAC Name 1H-1,3-benzodiazole-6-carbonitrile
SMILES N#CC1=CC=C2N=CNC2=C1

Specifications

Color Brown
Quantity 1 g
UN Number UN3439
Formula Weight 143.15
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Benzimidazole-6-carbonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.