Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzo[b]thiophene 97.0+%, TCI America™
SDP

Catalog No. B009325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B009325G 25 g
B00935G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B009325G Supplier TCI America Supplier No. B009325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 95-15-8
Molecular Formula C8H6S
Molecular Weight (g/mol) 134.196
MDL Number MFCD00005864
InChI Key FCEHBMOGCRZNNI-UHFFFAOYSA-N
Synonym benzo b thiophene, thianaphthene, benzothiophene, benzothiofuran, thionaphthene, 1-thiaindene, thianaphthen, benzothiophen, thianaphtene, 1-benzothiophen
PubChem CID 7221
ChEBI CHEBI:35858
IUPAC Name 1-benzothiophene
SMILES C1=CC=C2C(=C1)C=CS2

Specifications

Boiling Point 221°C
Quantity 25 g
UN Number 3077
Formula Weight 134.20
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Benzo[b]thiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.