Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Benzocyclobutene 94.0+%, TCI America™
SDP

Catalog No. B38631G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38631G 1 g
B3863200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B38631G Supplier TCI America Supplier No. B38631G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 694-87-1
Molecular Formula C8H8
Molecular Weight (g/mol) 104.152
MDL Number MFCD01321219
InChI Key UMIVXZPTRXBADB-UHFFFAOYSA-N
Synonym Bicyclo[4.2.0]octa-1,3,5-triene
PubChem CID 69667
ChEBI CHEBI:87328
IUPAC Name bicyclo[4.2.0]octa-1,3,5-triene
SMILES C1CC2=CC=CC=C21

Specifications

Color Yellow
Boiling Point 150°C
Quantity 1 g
UN Number 3295
Formula Weight 104.15
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzocyclobutene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.